Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238050.1 | XP_006238112.1 | 4 - 5 | 679 | 111 | |
XM_008763672.1 | XP_008761894.1 | 5 - 6 | 483 | 111 | ||
GenBank mRNA | FQ219084.1 | - | 4 - 5 | 631 | 111 | |
KJ817427.1 | - | 4 - 5 | 497 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |