Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173555.1 | NP_001167026.1 | 5 - 6 | 1173 | 82 | |
GenBank mRNA | BC079031.1 | - | 5 - 6 | 1173 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |