Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238141.2 | XP_006238203.1 | 9 - 10 | 2854 | 115 | |
XM_006238142.2 | XP_006238204.1 | 8 - 9 | 2023 | 115 | ||
XM_008763689.1 | XP_008761911.1 | 9 - 10 | 2683 | 115 | ||
XM_342829.7 | XP_342830.4 | 2633 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |