Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107919.1 | NP_001101389.1 | 7 - 8 | 1652 | 90 | |
XM_006237942.1 | XP_006238004.1 | 7 - 8 | 1659 | 90 | ||
GenBank mRNA | BC161859.1 | - | 7 - 8 | 1659 | 7096047 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |