Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080776.1 | NP_542954.1 | 5 - 6 | 1300 | 116 | |
XM_006237921.2 | XP_006237983.1 | 5 - 6 | 1366 | 116 | ||
GenBank mRNA | D63886.1 | - | 5 - 6 | 1300 | 116 | |
D85509.1 | - | 5 - 6 | 1300 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |