Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080776.1 | NP_542954.1 | 8 - 9 | 1801 | 79 | |
XM_006237921.2 | XP_006237983.1 | 8 - 9 | 1867 | 79 | ||
GenBank mRNA | D63886.1 | - | 8 - 9 | 1801 | 79 | |
D85509.1 | - | 8 - 9 | 1801 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |