Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138909.1 | NP_620264.1 | 1 | 97 | ||
GenBank mRNA | Y11321.1 | - | 1 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |