Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107921.1 | NP_001101391.1 | 6 - 7 | 672 | 122 | |
XM_006237973.2 | XP_006238035.1 | 7 - 8 | 572 | 122 | ||
XM_006237974.2 | XP_006238036.1 | 7 - 8 | 748 | 122 | ||
XM_006237975.2 | XP_006238037.1 | 6 - 7 | 502 | 122 | ||
XM_008763620.1 | XP_008761842.1 | 6 - 7 | 1666 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |