Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tstd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108663.1 | NP_001102133.1 | 6 - 7 | 1056 | 97 | |
XM_006238084.1 | XP_006238146.1 | 6 - 7 | 978 | 97 | ||
XM_006238085.2 | XP_006238147.1 | 6 - 7 | 1135 | 97 | ||
XM_006238086.1 | XP_006238148.1 | 5 - 6 | 871 | 97 | ||
XM_006238088.2 | XP_006238150.1 | 6 - 7 | 978 | 97 | ||
XM_006238089.2 | XP_006238151.1 | 6 - 7 | 978 | 97 | ||
XM_006238090.2 | XP_006238152.1 | 6 - 7 | 978 | 97 | ||
XM_006238092.2 | XP_006238154.1 | 6 - 7 | 978 | 97 | ||
XM_008763698.1 | XP_008761920.1 | 6 - 7 | 978 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons |