Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107917.2 | NP_001101387.2 | 7 - 8 | 737 | 141 | |
XM_006237932.1 | XP_006237994.1 | 8 - 9 | 859 | 141 | ||
GenBank mRNA | AM746508.1 | - | 7 - 8 | 737 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |