Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108457.1 | NP_001101927.1 | 5 - 6 | 487 | 158 | |
XM_006227621.2 | XP_006227683.1 | 5 - 6 | 521 | 158 | ||
GenBank mRNA | FQ225576.1 | - | 5 - 6 | 526 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |