Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276762.1 | NP_001263691.1 | 2 - 3 | 472 | 96 | |
GenBank mRNA | AF347030.1 | - | 460 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Rattus norvegicus Probe spans exons |