Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014249.1 | NP_001014271.1 | 2 - 3 | 652 | 90 | |
XM_008773004.1 | XP_008771226.1 | 2 - 3 | 724 | 90 | ||
GenBank mRNA | BC085898.1 | - | 2 - 3 | 652 | 7193369 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |