Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bves | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077590.1 | NP_001071058.1 | 6 - 7 | 977 | 136 | |
GenBank mRNA | AY786529.2 | - | 6 - 7 | 977 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |