Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014034.1 | NP_001014056.1 | 1 - 2 | 96 | 95 | |
XM_006256588.2 | XP_006256650.1 | 1 - 2 | 135 | 95 | ||
GenBank mRNA | BC089213.1 | - | 1 - 2 | 96 | 7302358 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |