Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113335.1 | NP_001106806.1 | 2 - 3 | 942 | 88 | |
NM_012653.2 | NP_036785.2 | 1 - 2 | 939 | 88 | ||
GenBank mRNA | L11004.1 | - | 1 - 2 | 954 | 88 | |
L11236.1 | - | 2 - 3 | 942 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |