Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080397.1 | NP_536322.1 | 1 - 2 | 221 | 84 | |
GenBank mRNA | AF022729.1 | - | 1 - 2 | 221 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |