Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012160.1 | NP_001012160.1 | 3 - 4 | 572 | 60 | |
XM_006253559.2 | XP_006253621.1 | 3 - 4 | 427 | 60 | ||
GenBank mRNA | BC083717.1 | - | 3 - 4 | 572 | 7190699 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |