Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112712.1 | NP_001106183.1 | 10 - 11 | 982 | 78 | |
NM_001112713.1 | NP_001106184.1 | 10 - 11 | 982 | 78 | ||
NM_031657.3 | NP_113845.2 | 10 - 11 | 982 | 78 | ||
XM_006253653.2 | XP_006253715.1 | 10 - 11 | 1091 | 78 | ||
XM_008771504.1 | XP_008769726.1 | 9 - 10 | 1249 | 78 | ||
GenBank mRNA | AF040750.1 | - | 10 - 11 | 982 | 78 | |
X97439.1 | - | 10 - 11 | 982 | 78 | ||
X97440.1 | - | 10 - 11 | 982 | 78 | ||
Y09365.1 | - | 10 - 11 | 1010 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |