Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013138.2 | NP_001013156.2 | 3 - 4 | 417 | 94 | |
XM_006253630.2 | XP_006253692.1 | 3 - 4 | 450 | 94 | ||
GenBank mRNA | AY485933.1 | - | 3 - 4 | 412 | 94 | |
BC097431.1 | - | 3 - 4 | 397 | 7455325 | 94 | |
FM038661.1 | - | 3 - 4 | 417 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |