Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109666.1 | NP_001103136.1 | 6 - 7 | 1104 | 69 | |
XM_006235929.2 | XP_006235991.1 | 7 - 8 | 1074 | 69 | ||
XM_006235930.2 | XP_006235992.1 | 7 - 8 | 1236 | 69 | ||
XM_006235931.2 | XP_006235993.1 | 7 - 8 | 1071 | 69 | ||
XM_006235932.2 | XP_006235994.1 | 7 - 8 | 1065 | 69 | ||
XM_006235933.2 | XP_006235995.1 | 7 - 8 | 1062 | 69 | ||
XM_006235934.2 | XP_006235996.1 | 7 - 8 | 1074 | 69 | ||
GenBank mRNA | BC129119.1 | - | 6 - 7 | 971 | 7115823 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |