Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109258.1 | NP_001102728.1 | 1 - 2 | 315 | 104 | |
GenBank mRNA | FQ213707.1 | - | 1 - 2 | 225 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |