Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107235.1 | NP_001100705.1 | 7 - 8 | 1202 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |