Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015022.3 | NP_001015022.1 | 2 - 3 | 315 | 114 | |
GenBank mRNA | AB210856.1 | - | 2 - 3 | 315 | 114 | |
BC093603.1 | - | 2 - 3 | 378 | 7316341 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |