Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015022.3 | NP_001015022.1 | 7 - 8 | 896 | 57 | |
GenBank mRNA | AB210856.1 | - | 7 - 8 | 896 | 57 | |
BC093603.1 | - | 7 - 8 | 959 | 7316341 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |