Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011912.1 | NP_001011912.1 | 10 - 11 | 1651 | 92 | |
XM_006251211.2 | XP_006251273.1 | 10 - 11 | 1866 | 92 | ||
XM_006251212.2 | XP_006251274.1 | 10 - 11 | 1854 | 92 | ||
XM_006251213.2 | XP_006251275.1 | 10 - 11 | 2032 | 92 | ||
XM_006251214.2 | XP_006251276.1 | 10 - 11 | 1274 | 92 | ||
GenBank mRNA | BC081868.1 | - | 10 - 11 | 1651 | 7096894 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |