Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270849.1 | NP_001257778.1 | 2 - 3 | 303 | 76 | |
GenBank mRNA | X53589.1 | - | 2 - 3 | 303 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |