Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126275.1 | NP_001119747.1 | 3 - 4 | 325 | 116 | |
XM_006251318.2 | XP_006251380.1 | 3 - 4 | 2169 | 116 | ||
XM_006251319.2 | XP_006251381.1 | 3 - 4 | 610 | 116 | ||
GenBank mRNA | BC160889.1 | - | 3 - 4 | 325 | 7441909 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |