Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181365.2 | NP_852030.1 | 668 | 96 | ||
GenBank mRNA | AF345444.1 | - | 668 | 96 | ||
AF345445.1 | - | 1 - 2 | 238 | 96 | ||
AF453245.1 | - | 2 - 3 | 291 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |