Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105750.2 | NP_001099220.2 | 2 - 3 | 488 | 97 | |
XM_008770187.1 | XP_008768409.1 | 2 - 3 | 7130 | 97 | ||
GenBank mRNA | FQ231655.1 | - | 3 - 4 | 220 | 97 | |
FQ235297.1 | - | 3 - 4 | 220 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |