Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nacad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC091311.1 | - | 4 - 5 | 1485 | 70 | |
BI289520.1 | - | 3 - 4 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |