Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108355.1 | NP_001101825.1 | 9 - 10 | 1157 | 93 | |
XM_006250708.2 | XP_006250770.1 | 9 - 10 | 1189 | 93 | ||
XM_006250709.2 | XP_006250771.1 | 9 - 10 | 1076 | 93 | ||
XM_008770042.1 | XP_008768264.1 | 8 - 9 | 3956 | 93 | ||
XM_008770043.1 | XP_008768265.1 | 8 - 9 | 968 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |