Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspbap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134419.2 | NP_599246.2 | 4 - 5 | 625 | 154 | |
GenBank mRNA | AF168362.1 | - | 4 - 5 | 614 | 154 | |
BC091125.1 | - | 4 - 5 | 625 | 7366096 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |