Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047846.1 | NP_001041311.1 | 3 - 4 | 353 | 66 | |
XM_006248241.2 | XP_006248303.1 | 2 - 3 | 326 | 66 | ||
XM_008768630.1 | XP_008766852.1 | 2 - 3 | 385 | 66 | ||
GenBank mRNA | AY325209.1 | - | 3 - 4 | 353 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |