Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053540.2 | NP_445992.1 | 2 - 3 | 293 | 116 | |
GenBank mRNA | BI293817.1 | - | 1 - 2 | 116 | ||
BQ195675.1 | - | 1 - 2 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |