Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105879.1 | NP_001099349.1 | 10 - 11 | 1839 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |