Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024899.1 | NP_001020070.1 | 1 - 2 | 352 | 84 | |
XM_006231546.2 | XP_006231608.1 | 1 - 2 | 406 | 84 | ||
GenBank mRNA | BC097447.1 | - | 1 - 2 | 352 | 7456556 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |