Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024899.1 | NP_001020070.1 | 4 - 5 | 761 | 88 | |
XM_006231546.2 | XP_006231608.1 | 4 - 5 | 815 | 88 | ||
XM_008760443.1 | XP_008758665.1 | 3 - 4 | 821 | 88 | ||
GenBank mRNA | BC097447.1 | - | 4 - 5 | 761 | 7456556 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |