Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231570.2 | XP_006231632.1 | 4 - 5 | 2378 | 86 | |
XM_008760486.1 | XP_008758708.1 | 4 - 5 | 2887 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |