Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243251.2 | XP_006243313.1 | 8 - 9 | 1056 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |