Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022536.2 | NP_071981.1 | 4 - 5 | 527 | 144 | |
GenBank mRNA | AF071225.1 | - | 4 - 5 | 527 | 144 | |
BC061971.1 | - | 4 - 5 | 556 | 5623342 | 144 | |
FQ211358.1 | - | 4 - 5 | 531 | 144 | ||
FQ212031.1 | - | 4 - 5 | 556 | 144 | ||
FQ219792.1 | - | 4 - 5 | 565 | 144 | ||
FQ219926.1 | - | 4 - 5 | 577 | 144 | ||
FQ221020.1 | - | 4 - 5 | 580 | 144 | ||
FQ221362.1 | - | 4 - 5 | 554 | 144 | ||
FQ222696.1 | - | 4 - 5 | 689 | 144 | ||
FQ223162.1 | - | 4 - 5 | 574 | 144 | ||
FQ228087.1 | - | 4 - 5 | 570 | 144 | ||
FQ229174.1 | - | 4 - 5 | 577 | 144 | ||
FQ229214.1 | - | 4 - 5 | 571 | 144 | ||
FQ230004.1 | - | 581 | 144 | |||
FQ231656.1 | - | 4 - 5 | 570 | 144 | ||
FQ232077.1 | - | 4 - 5 | 566 | 144 | ||
FQ232920.1 | - | 4 - 5 | 580 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |