Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017075.2 | NP_058771.2 | 8 - 9 | 905 | 138 | |
GenBank mRNA | D00512.1 | - | 8 - 9 | 837 | 138 | |
D13921.1 | - | 8 - 9 | 837 | 138 | ||
FM098848.1 | - | 3 - 4 | 287 | 138 | ||
FQ217320.1 | - | 8 - 9 | 843 | 138 | ||
FQ228108.1 | - | 8 - 9 | 891 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |