Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127554.1 | NP_001121026.1 | 2 - 3 | 412 | 89 | |
GenBank mRNA | BC166915.1 | - | 2 - 3 | 412 | 6917859 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |