Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hemk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106853.1 | NP_001100323.1 | 2 - 3 | 418 | 92 | |
XM_006243728.2 | XP_006243790.1 | 3 - 4 | 708 | 92 | ||
XM_006243729.2 | XP_006243791.1 | 3 - 4 | 1028 | 92 | ||
XM_006243730.2 | XP_006243792.1 | 2 - 3 | 783 | 92 | ||
XM_008766502.1 | XP_008764724.1 | 3 - 4 | 643 | 92 | ||
GenBank mRNA | BC161944.1 | - | 2 - 3 | 412 | 9024451 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |