Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012600.2 | NP_036732.2 | 7 - 8 | 818 | 88 | |
XM_008766439.1 | XP_008764661.1 | 7 - 8 | 1115 | 88 | ||
GenBank mRNA | CO560778.1 | - | 3 - 4 | 315 | 88 | |
FQ228526.1 | - | 7 - 8 | 849 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |