Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133427.3 | NP_596918.3 | 13 - 14 | 1396 | 118 | |
XM_006243472.2 | XP_006243534.1 | 12 - 13 | 1382 | 118 | ||
XM_008766437.1 | XP_008764659.1 | 13 - 14 | 1555 | 118 | ||
GenBank mRNA | AF307840.1 | - | 12 - 13 | 1155 | 118 | |
AY321370.1 | - | 12 - 13 | 1155 | 118 | ||
AY321371.1 | - | 11 - 12 | 1002 | 118 | ||
BC080240.1 | - | 13 - 14 | 1374 | 7099950 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |