Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005548.1 | NP_001005548.1 | 9 - 10 | 1664 | 68 | |
GenBank mRNA | BC082098.1 | - | 9 - 10 | 1664 | 7134452 | 68 |
FQ229860.1 | - | 9 - 10 | 1664 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |