Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163477.1 | NP_001156949.1 | 1 - 2 | 307 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |