Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109004.2 | NP_001102474.1 | 8 - 9 | 787 | 85 | |
XM_008766558.1 | XP_008764780.1 | 8 - 9 | 1396 | 85 | ||
GenBank mRNA | BC162006.1 | - | 8 - 9 | 787 | 9089897 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |