Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170539.1 | NP_001164010.1 | 2 - 3 | 481 | 76 | |
GenBank mRNA | AY310157.1 | - | 216 | 76 | ||
BC166740.1 | - | 2 - 3 | 481 | 7937421 | 76 | |
FQ210494.1 | - | 2 - 3 | 482 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |